accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

FFFSOOHOFFFSOOHOSn

CAS RN 62086-04-8

IUPAC Name: bis(trifluoromethanesulfonic acid) tin
Synonyms: bis(trifluoromethanesulfonic acid) tin
Molecular Weight (g/mol): 418.85
Molecular Formula: C2H2F6O6S2Sn
InChi Key: HKYRUGOZNFKLHI-UHFFFAOYSA-N
SMILES: [Sn].OS(=O)(=O)C(F)(F)F.OS(=O)(=O)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results