CAS RN 62096-63-3
IUPAC Name:
6-ethoxy-N2-methyl-1,3,5-triazine-2,4-diamine
Synonyms:
6-ethoxy-N2-methyl-1,3,5-triazine-2,4-diamine
Molecular Weight (g/mol):
169.19
Molecular Formula:
C6H11N5O
InChi Key:
HEMSJLNVJJUYEU-UHFFFAOYSA-N
SMILES:
CCOC1=NC(N)=NC(NC)=N1