accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 621-23-8

IUPAC Name: 1,3,5-trimethoxybenzene
Synonyms: 1,3,5-trimethoxybenzene 1,3,5-trimethyoxybenzene sym-trimethoxybenzene
Molecular Weight (g/mol): 168.19
Molecular Formula: C9H12O3
InChi Key: LKUDPHPHKOZXCD-UHFFFAOYSA-N
SMILES: COC1=CC(OC)=CC(OC)=C1
Quantity 
  • (1)
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results