accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 621-79-4

IUPAC Name: (2E)-3-phenylprop-2-enamide
Synonyms: cinnamamide
Molecular Weight (g/mol): 147.18
Molecular Formula: C9H9NO
InChi Key: APEJMQOBVMLION-VOTSOKGWSA-N
SMILES: NC(=O)\C=C\C1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (2)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results