accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3H3CCH3OONH(S)BrHOO

CAS RN 62129-39-9

IUPAC Name: (2S)-3-(4-bromophenyl)-2-{[(tert-butoxy)carbonyl]amino}propanoate
Synonyms: (2S)-3-(4-bromophenyl)-2-[(tert-butoxycarbonyl)amino]propanoate
Molecular Weight (g/mol): 343.2
Molecular Formula: C14H17BrNO4
InChi Key: ULNOXUAEIPUJMK-NSHDSACASA-M
SMILES: CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(Br)C=C1)C([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results