accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NOOOSOOOK

CAS RN 6217-68-1

IUPAC Name: potassium 4-nitrophenyl sulfate
Synonyms: potassium p-nitrophenyl sulfate
Molecular Weight (g/mol): 257.26
Molecular Formula: C6H4KNO6S
InChi Key: BITVAZYUWRLLCN-UHFFFAOYSA-M
SMILES: [K+].[O-][N+](=O)C1=CC=C(OS([O-])(=O)=O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)