accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-20-8

IUPAC Name: {[2-(phenylsulfanyl)ethyl]sulfanyl}benzene
Synonyms: {[2-(phenylsulfanyl)ethyl]sulfanyl}benzene
Molecular Weight (g/mol): 246.39
Molecular Formula: C14H14S2
InChi Key: MHCVYAFXPIMYRD-UHFFFAOYSA-N
SMILES: C(CSC1=CC=CC=C1)SC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results