accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-26-4

OHNH2

CAS RN 622-26-4

IUPAC Name: 4-(2-hydroxyethyl)piperidin-1-ium
Synonyms: 4-(2-hydroxyethyl)piperidin-1-ium
Molecular Weight (g/mol): 130.21
Molecular Formula: C7H16NO
InChi Key: LDSQQXKSEFZAPE-UHFFFAOYSA-O
SMILES: OCCC1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)