accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-29-7

CH3N(E/Z)

CAS RN 622-29-7

IUPAC Name: methyl(phenylmethylidene)amine
Synonyms: methyl(phenylmethylidene)amine
Molecular Weight (g/mol): 119.17
Molecular Formula: C8H9N
InChi Key: HXTGGPKOEKKUQO-UHFFFAOYSA-N
SMILES: CN=CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (2)
Quantity 
  • (2)
  • (2)
  • (2)
Boiling Point 
  • (3)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (4)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)