accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3S

CAS RN 622-38-8

IUPAC Name: (ethylsulfanyl)benzene
Synonyms: phenyl ethyl sulfide
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: AEHWKBXBXYNPCX-UHFFFAOYSA-N
SMILES: CCSC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (2)
  • (2)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results