accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NCO

CAS RN 622-58-2

IUPAC Name: 1-isocyanato-4-methylbenzene
Synonyms: p-tolyl isocyanate 4-Tol-NCO p-tmi PTI TMI
Molecular Weight (g/mol): 133.15
Molecular Formula: C8H7NO
InChi Key: MGYGFNQQGAQEON-UHFFFAOYSA-N
SMILES: CC1=CC=C(C=C1)N=C=O
Boiling Point 
  • (3)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (4)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (4)