accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-76-4

H3C

CAS RN 622-76-4

IUPAC Name: (but-1-yn-1-yl)benzene
Synonyms: but-1-yn-1-ylbenzene
Molecular Weight (g/mol): 130.19
Molecular Formula: C10H10
InChi Key: FFFMSANAQQVUJA-UHFFFAOYSA-N
SMILES: CCC#CC1=CC=CC=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)