accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 622-88-8

HClH2NNHBr

CAS RN 622-88-8

IUPAC Name: hydrogen (4-bromophenyl)hydrazine chloride
Synonyms: hydrogen hydrazine, (4-bromophenyl)- chloride
Molecular Weight (g/mol): 223.5
Molecular Formula: C6H8BrClN2
InChi Key: RGGOWBBBHWTTRE-UHFFFAOYSA-N
SMILES: [H+].[Cl-].NNC1=CC=C(Br)C=C1
Molecular Weight (g/mol) 
  • (2)
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)