accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CCH3

CAS RN 622-96-8

IUPAC Name: 1-ethyl-4-methylbenzene
Synonyms: 4-ethyltoluene P-ethyltoluene
Molecular Weight (g/mol): 120.2
Molecular Formula: C9H12
InChi Key: JRLPEMVDPFPYPJ-UHFFFAOYSA-N
SMILES: CCC1=CC=C(C)C=C1
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
Solvent or Matrix 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results