accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62211-93-2

IUPAC Name: 4,5-bis(acetyloxy)-2-methyloxolan-3-yl acetate
Synonyms: 4,5-bis(acetyloxy)-2-methyloxolan-3-yl acetate
Molecular Weight (g/mol): 260.24
Molecular Formula: C11H16O7
InChi Key: NXEJETQVUQAKTO-UHFFFAOYNA-N
SMILES: CC1OC(OC(C)=O)C(OC(C)=O)C1OC(C)=O
Quantity 
  • (2)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results