accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3NNOH

CAS RN 622381-65-1

IUPAC Name: {4-[(4-methylpiperazin-1-yl)methyl]phenyl}methanol
Synonyms: {4-[(4-methylpiperazin-1-yl)methyl]phenyl}methanol
Molecular Weight (g/mol): 220.32
Molecular Formula: C13H20N2O
InChi Key: ADMFZWUKZBAYIJ-UHFFFAOYSA-N
SMILES: CN1CCN(CC2=CC=C(CO)C=C2)CC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results