CAS RN 6226-79-5
Ponceau S, Electrophoresis Grade
CAS: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Molecular Weight (g/mol): 760.552 MDL Number: MFCD00003892 InChI Key: VSXKEUCERCWGKF-STNZDNLRSA-J Synonym: C.I. 27195 PubChem CID: 11320219 IUPAC Name: tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Ponceau S, 0.1% v/v soln. in 5% acetic acid
CAS: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Molecular Weight (g/mol): 760.552 MDL Number: MFCD00003892 InChI Key: VSXKEUCERCWGKF-STNZDNLRSA-J Synonym: ponceau s,acid red 112,ponceau s extra,ponceau red s,unii-t1c2ugs05f,c.i. acid red 112,t1c2ugs05f,3-hydroxy-4-2-sulfo-4-4-sulfophenylazo phenylazo-2,7-naphthalenedisulfonic acid sodium salt,c.i. acid red 112, tetrasodium salt,ponceau-s PubChem CID: 11320219 IUPAC Name: tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Thermo Scientific Chemicals Ponceau S, pure
CAS: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Molecular Weight (g/mol): 760.552 MDL Number: MFCD00003892 InChI Key: VSXKEUCERCWGKF-STNZDNLRSA-J Synonym: C.I. 27195,3-Hydroxy-4-[2-sulfo-4-(4-sulfophenylazo)phenylazo]-2, 7-naphthalenedisulfonic acid, tetrasodium salt PubChem CID: 11320219 IUPAC Name: tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Ponceau S, 85%, MP Biomedicals Inc
CAS: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Molecular Weight (g/mol): 760.552 InChI Key: VSXKEUCERCWGKF-STNZDNLRSA-J Synonym: 3-Hydroxy-4-(2-sulfo-4-(4-sulfophenylazo)phenylazo)-2, 7-naphthalene-disulfonic acid tetrasodium salt,C.I. 27195 PubChem CID: 11320219 IUPAC Name: tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]
Ponceau S, MP Biomedicals
CAS: 6226-79-5 Molecular Formula: C22H12N4Na4O13S4 Molecular Weight (g/mol): 760.552 InChI Key: VSXKEUCERCWGKF-STNZDNLRSA-J Synonym: 3-Hydroxy-4-[2-sulfo-4-(4-sulfo- phenylazo)phenylazo]-2, 7- naphthalene-disulfonic acid tetrasodium salt PubChem CID: 11320219 IUPAC Name: tetrasodium;(4Z)-3-oxo-4-[[2-sulfonato-4-[(4-sulfonatophenyl)diazenyl]phenyl]hydrazinylidene]naphthalene-2,7-disulfonate SMILES: C1=CC(=CC=C1N=NC2=CC(=C(C=C2)NN=C3C4=C(C=C(C=C4)S(=O)(=O)[O-])C=C(C3=O)S(=O)(=O)[O-])S(=O)(=O)[O-])S(=O)(=O)[O-].[Na+].[Na+].[Na+].[Na+]