accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3CSCH3

CAS RN 623-13-2

IUPAC Name: 1-methyl-4-(methylsulfanyl)benzene
Synonyms: 1-methyl-4-(methylsulfanyl)benzene
Molecular Weight (g/mol): 138.23
Molecular Formula: C8H10S
InChi Key: VHILIAIEEYLJNA-UHFFFAOYSA-N
SMILES: CSC1=CC=C(C)C=C1
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)