accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 623-27-8

OO

CAS RN 623-27-8

IUPAC Name: benzene-1,4-dicarbaldehyde
Synonyms: terephthalaldehyde
Molecular Weight (g/mol): 134.13
Molecular Formula: C8H6O2
InChi Key: KUCOHFSKRZZVRO-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(C=O)C=C1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (2)
Boiling Point 
  • (3)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (2)