accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClOON

CAS RN 62348-13-4

IUPAC Name: 1,2-oxazole-5-carbonyl chloride
Synonyms: 1,2-oxazole-5-carbonyl chloride
Molecular Weight (g/mol): 131.52
Molecular Formula: C4H2ClNO2
InChi Key: NASLINFISOTVJJ-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC=NO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results