accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62348-24-7

IUPAC Name: 4-methyl-1,3-oxazole-5-carbonyl chloride
Synonyms: 4-methyl-1,3-oxazole-5-carbonyl chloride
Molecular Weight (g/mol): 145.54
Molecular Formula: C5H4ClNO2
InChi Key: YPKNOSGIABPXKS-UHFFFAOYSA-N
SMILES: CC1=C(OC=N1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results