accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H3COHOHO

CAS RN 624-00-0

IUPAC Name: 5-hydroxydecanoic acid
Synonyms: 5-hydroxy capric acid
Molecular Weight (g/mol): 188.27
Molecular Formula: C10H20O3
InChi Key: LMHJFKYQYDSOQO-UHFFFAOYNA-N
SMILES: CCCCCC(O)CCCC(O)=O
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results