accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HHClClNH2NH2

CAS RN 624-18-0

IUPAC Name: dihydrogen benzene-1,4-diamine dichloride
Synonyms: P-phenylenediamine hcl
Molecular Weight (g/mol): 181.06
Molecular Formula: C6H10Cl2N2
InChi Key: IYXMNTLBLQNMLM-UHFFFAOYSA-N
SMILES: [H+].[H+].[Cl-].[Cl-].NC1=CC=C(N)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)