accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 624-38-4

IUPAC Name: 1,4-diiodobenzene
Synonyms: 1,4-diiodobenzene
Molecular Weight (g/mol): 329.91
Molecular Formula: C6H4I2
InChi Key: LFMWZTSOMGDDJU-UHFFFAOYSA-N
SMILES: IC1=CC=C(I)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results