accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 624-39-5

IUPAC Name: (4-sulfanidylphenyl)sulfanide
Synonyms: (4-sulfanidylphenyl)sulfanide
Molecular Weight (g/mol): 140.22
Molecular Formula: C6H4S2
InChi Key: WYLQRHZSKIDFEP-UHFFFAOYSA-L
SMILES: [S-]C1=CC=C([S-])C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results