accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6245-57-4

IUPAC Name: 4-methoxy-2-methylbenzoic acid
Synonyms: 4-methoxy-2-methylbenzoic acid
Molecular Weight (g/mol): 166.18
Molecular Formula: C9H10O3
InChi Key: MSVRGYOYISBGTH-UHFFFAOYSA-N
SMILES: COC1=CC(C)=C(C=C1)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results