accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CH3OHH2C

CAS RN 625-31-0

IUPAC Name: pent-4-en-2-ol
Synonyms: 4-penten-2-ol
Molecular Weight (g/mol): 86.13
Molecular Formula: C5H10O
InChi Key: ZHZCYWWNFQUZOR-UHFFFAOYNA-N
SMILES: CC(O)CC=C
Boiling Point 
  • (3)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)