accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClClO

CAS RN 625-36-5

IUPAC Name: 3-chloropropanoyl chloride
Synonyms: propanoyl chloride, 3-chloro-
Molecular Weight (g/mol): 126.96
Molecular Formula: C3H4Cl2O
InChi Key: INUNLMUAPJVRME-UHFFFAOYSA-N
SMILES: ClCCC(Cl)=O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)