accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OHNOO

CAS RN 625-48-9

IUPAC Name: 2-nitroethan-1-ol
Synonyms: 2-nitroethanol
Molecular Weight (g/mol): 91.07
Molecular Formula: C2H5NO3
InChi Key: KIPMDPDAFINLIV-UHFFFAOYSA-N
SMILES: OCC[N+]([O-])=O
Molecular Weight (g/mol) 
  • (3)
  • (6)
Quantity 
  • (1)
  • (2)
  • (3)
  • (3)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (6)
  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)