CAS RN 6250-70-0
CAS RN 6250-70-0
IUPAC Name:
nonadecanedioic acid
Synonyms:
nonadecanedioic acid
Molecular Weight (g/mol):
328.49
Molecular Formula:
C19H36O4
InChi Key:
IFAWYXIHOVRGHQ-UHFFFAOYSA-N
SMILES:
OC(=O)CCCCCCCCCCCCCCCCCC(O)=O
1
–
1
of
1
results
1
Nonadecanedioic Acid, TRC
CAS: 6250-70-0 Molecular Formula: C19H36O4 Molecular Weight (g/mol): 328.49 IUPAC Name: nonadecanedioic acid SMILES: OC(=O)CCCCCCCCCCCCCCCCCC(=O)O