accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6258-60-2

IUPAC Name: (4-methoxyphenyl)methanethiol
Synonyms: (4-methoxyphenyl)methanethiol
Molecular Weight (g/mol): 154.23
Molecular Formula: C8H10OS
InChi Key: PTDVPWWJRCOIIO-UHFFFAOYSA-N
SMILES: COC1=CC=C(CS)C=C1
Boiling Point 
  • (4)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results