accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

HSS

CAS RN 6258-63-5

IUPAC Name: (thiophen-2-yl)methanethiol
Synonyms: thiophen-2-ylmethanethiol
Molecular Weight (g/mol): 130.22
Molecular Formula: C5H6S2
InChi Key: GCZQHDFWKVMZOE-UHFFFAOYSA-N
SMILES: SCC1=CC=CS1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results