accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 626-02-8

IUPAC Name: 3-iodophenol
Synonyms: 3-iodophenol m-jodphenol 3-jodphenol
Molecular Weight (g/mol): 220.01
Molecular Formula: C6H5IO
InChi Key: FXTKWBZFNQHAAO-UHFFFAOYSA-N
SMILES: OC1=CC=CC(I)=C1
Molecular Weight (g/mol) 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (3)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (3)
  • (2)
  • (2)