accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 626-05-1

BrBrN

CAS RN 626-05-1

IUPAC Name: 2,6-dibromopyridine
Synonyms: pyridine, 2,6-dibromo-
Molecular Weight (g/mol): 236.89
Molecular Formula: C5H3Br2N
InChi Key: FEYDZHNIIMENOB-UHFFFAOYSA-N
SMILES: BrC1=CC=CC(Br)=N1
Molecular Weight (g/mol) 
  • (6)
  • (4)
Quantity 
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
Percent Purity 
  • (1)
  • (6)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (1)
  • (3)
  • (6)
  • (4)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (2)
  • (1)
  • (6)
  • (3)
  • (3)