accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

NH2BrBr

CAS RN 626-40-4

IUPAC Name: 3,5-dibromoaniline
Synonyms: 3,5-dibromoaniline
Molecular Weight (g/mol): 250.92
Molecular Formula: C6H5Br2N
InChi Key: RVNUUWJGSOHMRR-UHFFFAOYSA-N
SMILES: NC1=CC(Br)=CC(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results