accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

ClN

CAS RN 626-60-8

IUPAC Name: 3-chloropyridine
Synonyms: 3-chloropyridine
Molecular Weight (g/mol): 113.54
Molecular Formula: C5H4ClN
InChi Key: PWRBCZZQRRPXAB-UHFFFAOYSA-N
SMILES: ClC1=CC=CN=C1
Boiling Point 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results