accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 626-64-2

ONH

CAS RN 626-64-2

IUPAC Name: 1,4-dihydropyridin-4-one
Synonyms: 4pyridone 4-pyridone
Molecular Weight (g/mol): 95.1
Molecular Formula: C5H5NO
InChi Key: GCNTZFIIOFTKIY-UHFFFAOYSA-N
SMILES: O=C1C=CNC=C1
Molecular Weight (g/mol) 
  • (1)
  • (6)
Quantity 
  • (2)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (1)
  • (1)
  • (6)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)