accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 626-93-7

H3CCH3OH

CAS RN 626-93-7

IUPAC Name: hexan-2-ol
Synonyms: 2-hexanol
Molecular Weight (g/mol): 102.18
Molecular Formula: C6H14O
InChi Key: QNVRIHYSUZMSGM-UHFFFAOYNA-N
SMILES: CCCCC(C)O
Boiling Point 
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)