accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 626-99-3

OHOCH2(E)

CAS RN 626-99-3

IUPAC Name: (2E)-penta-2,4-dienoic acid
Synonyms: (E)-2,4-pentadienoic acid β-vinyl acrylic acid
Molecular Weight (g/mol): 98.1
Molecular Formula: C5H6O2
InChi Key: SDVVLIIVFBKBMG-ONEGZZNKSA-N
SMILES: OC(=O)\C=C\C=C
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results