accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

KOO

CAS RN 62638-03-3

IUPAC Name: potassium 4-cyclohexylbutanoate
Synonyms: potassium 4-cyclohexylbutanoate
Molecular Weight (g/mol): 208.34
Molecular Formula: C10H17KO2
InChi Key: LBYNWSSEILNGLN-UHFFFAOYSA-M
SMILES: [K+].[O-]C(=O)CCCC1CCCCC1
Quantity 
  • (2)
  • (1)
  • (2)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (5)
  • (2)
  • (1)
  • (2)
  • (2)
  • (2)