accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OONHO

CAS RN 62642-62-0

IUPAC Name: 4-[(4-carboxylatophenyl)methyl]morpholin-4-ium
Synonyms: 4-[(4-carboxylatophenyl)methyl]morpholin-4-ium
Molecular Weight (g/mol): 221.26
Molecular Formula: C12H15NO3
InChi Key: QYBXZYYECZFQRX-UHFFFAOYSA-N
SMILES: [O-]C(=O)C1=CC=C(C[NH+]2CCOCC2)C=C1
Molecular Weight (g/mol) 
  • (5)
  • (2)
Quantity 
  • (3)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results