accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6269-33-6

IUPAC Name: (2E)-4-(4-chlorophenyl)-4-oxobut-2-enoic acid
Synonyms: (2E)-4-(4-chlorophenyl)-4-oxobut-2-enoic acid
Molecular Weight (g/mol): 210.61
Molecular Formula: C10H7ClO3
InChi Key: VQVQEUFKSRHRCT-AATRIKPKSA-N
SMILES: OC(=O)\C=C\C(=O)C1=CC=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results