accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 6269-89-2

IUPAC Name: 1-(4-nitrophenyl)piperazine
Synonyms: 1-(4-nitrophenyl)piperazine
Molecular Weight (g/mol): 207.23
Molecular Formula: C10H13N3O2
InChi Key: VWOJSRICSKDKAW-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC=C(C=C1)N1CCNCC1
Molecular Weight (g/mol) 
  • (3)
  • (3)
Percent Purity 
  • (3)
  • (3)
Quantity 
  • (2)
  • (2)
  • (2)
Physical Form 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (2)
  • (2)
  • (3)