accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

II

CAS RN 627-31-6

IUPAC Name: 1,3-diiodopropane
Synonyms: propane, 1,3-diiodo-
Molecular Weight (g/mol): 295.89
Molecular Formula: C3H6I2
InChi Key: AAAXMNYUNVCMCJ-UHFFFAOYSA-N
SMILES: ICCCI
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)