accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 627-75-8

IUPAC Name: hydrogen (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Synonyms: hydrogen (2R)-2-amino-5-[(diaminomethylidene)amino]pentanoic acid chloride
Molecular Weight (g/mol): 210.66
Molecular Formula: C6H15ClN4O2
InChi Key: KWTQSFXGGICVPE-PGMHMLKASA-N
SMILES: [H+].[Cl-].N[C@H](CCCN=C(N)N)C(O)=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)