accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

OO(R)OO(S)OOOO

CAS RN 62708-56-9

IUPAC Name: (2R,3S)-2,3-bis(benzoyloxy)butanedioate
Synonyms: (2R,3S)-2,3-bis(benzoyloxy)butanedioate
Molecular Weight (g/mol): 356.29
Molecular Formula: C18H12O8
InChi Key: YONLFQNRGZXBBF-OKILXGFUSA-L
SMILES: [O-]C(=O)[C@@H](OC(=O)C1=CC=CC=C1)[C@@H](OC(=O)C1=CC=CC=C1)C([O-])=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results