accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62720-29-0

IUPAC Name: 1,3-dibromo-5-fluoro-2-iodobenzene
Synonyms: 1,3-dibromo-5-fluoro-2-iodobenzene
Molecular Weight (g/mol): 379.79
Molecular Formula: C6H2Br2FI
InChi Key: LIWKANDEJFABTQ-UHFFFAOYSA-N
SMILES: FC1=CC(Br)=C(I)C(Br)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results