accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62790-50-5

IUPAC Name: 4-chloro-2-methyl-6-nitroaniline
Synonyms: 4-chloro-2-methyl-6-nitroaniline
Molecular Weight (g/mol): 186.6
Molecular Formula: C7H7ClN2O2
InChi Key: QDSCDFKGUAONPC-UHFFFAOYSA-N
SMILES: CC1=CC(Cl)=CC(=C1N)[N+]([O-])=O
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results