accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

CAS RN 62796-29-6

CH3NCH3OSOOOSClOONH3CH3C(?)

CAS RN 62796-29-6

IUPAC Name: 9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Synonyms: 9-[4-(chlorosulfonyl)-2-sulfonatophenyl]-3,6-bis(diethylamino)-10λ⁴-xanthen-10-ylium
Molecular Weight (g/mol): 577.11
Molecular Formula: C27H29ClN2O6S2
InChi Key: YERWMQJEYUIJBO-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC2=[O+]C3=CC(=CC=C3C(C3=CC=C(C=C3S([O-])(=O)=O)S(Cl)(=O)=O)=C2C=C1)N(CC)CC

 No Refinements