accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Français.

H2NONH2O

CAS RN 628-94-4

IUPAC Name: hexanediamide
Synonyms: adipamide
Molecular Weight (g/mol): 144.17
Molecular Formula: C6H12N2O2
InChi Key: GVNWZKBFMFUVNX-UHFFFAOYSA-N
SMILES: NC(=O)CCCCC(N)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results